siRNA Details
Gene Symbol: 9130008F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027834.3 | NP_082110.1 | 2 | 1139 | ||
GenBank mRNA | AK018601.1 | - | 2 | 1140 | ||
AK033670.1 | - | 2 | 1135 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9130008F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027834.3 | NP_082110.1 | 2 | 1139 | ||
GenBank mRNA | AK018601.1 | - | 2 | 1140 | ||
AK033670.1 | - | 2 | 1135 |