siRNA Details
| Gene Symbol: 9130008F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027834.3 | NP_082110.1 | 2 | 1978 | ||
| GenBank mRNA | AK033670.1 | - | 2 | 1974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130008F23Rik| Gene Symbol: 9130008F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027834.3 | NP_082110.1 | 2 | 1978 | ||
| GenBank mRNA | AK033670.1 | - | 2 | 1974 | ||