siRNA Details
| Gene Symbol: C2cd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027909.2 | NP_082185.2 | 13 | 2083 | ||
| GenBank mRNA | AK004920.1 | - | 13 | 2088 | ||
| AK039220.1 | - | 13 | 2054 | |||
| AK122237.1 | - | 8 | 3658 | |||
| AK153804.1 | - | 13 | 2055 | |||
| BC035271.1 | - | 10 | 2089 | |||
| BC049875.1 | - | 9 | 1010 | |||
| BC056612.1 | - | 13 | 1654 | 4952570 | ||
| BC060156.1 | - | 13 | 1947 | 6810220 | ||

C2cd2l
Genomic Map