siRNA Details
| Gene Symbol: Dcaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027946.3 | NP_082222.1 | 7 | 1705 | ||
| GenBank mRNA | AK047435.1 | - | 7 | 1707 | ||
| AK154757.1 | - | 7 | 1696 | |||
| AK190173.1 | - | 1 | 150 | |||
| BC048165.1 | - | 7 | 1618 | 6409531 | ||

Dcaf7
Genomic Map