siRNA Details
| Gene Symbol: Dcaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027946.3 | NP_082222.1 | 6 | 1032 | ||
| GenBank mRNA | AK005902.1 | - | 6 | 1026 | ||
| AK047435.1 | - | 6 | 1034 | |||
| AK154757.1 | - | 6 | 1023 | |||
| AK165674.1 | - | 6 | 1023 | |||
| BC048165.1 | - | 6 | 945 | 6409531 | ||
| BC048722.1 | - | 6 | 951 | 6504571 | ||

Dcaf7
Genomic Map