siRNA Details
| Gene Symbol: AIM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039775.3 | NP_001034864.2 | 2 | 314 | ||
| XM_005245918.2 | XP_005245975.1 | 2 | 300 | |||
| XM_011541672.1 | XP_011539974.1 | 2 | 300 | |||
| XM_011541673.2 | XP_011539975.1 | 2 | 539 | |||
| GenBank mRNA | AK000902.1 | - | 2 | 314 | ||
| DA439029.1 | - | 2 | 300 | |||

AIM1L
Genomic Map