siRNA Details
| Gene Symbol: RNF157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052916.2 | NP_443148.1 | 19 | 3348 | ||
| XM_005257007.4 | XP_005257064.1 | 20 | 3448 | |||
| XM_017024119.1 | XP_016879608.1 | 18 | 3341 | |||
| GenBank mRNA | AB067504.2 | - | 19 | 3349 | ||
| AK055949.1 | - | 10 | 2428 | |||
| BC004231.2 | - | 9 | 1185 | |||
| CX872674.1 | - | 1 | 158 | |||

RNF157
Genomic Map