siRNA Details
Gene Symbol: Wbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053766.1 | NP_446218.1 | 9 | 927 | ||
XM_008770840.1 | XP_008769062.1 | 6 | 1443 | |||
XM_008770841.1 | XP_008769063.1 | 7 | 946 | |||
GenBank mRNA | BC089052.1 | - | 9 | 927 | 7307299 | |
FQ226684.1 | - | 8 | 709 |
