siRNA Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053766.1 | NP_446218.1 | 11 | 1250 | ||
| XM_008770840.1 | XP_008769062.1 | 8 | 1766 | |||
| XM_008770841.1 | XP_008769063.1 | 9 | 1269 | |||
| GenBank mRNA | BC089052.1 | - | 11 | 1250 | 7307299 | |

Wbp4
Genomic Map