siRNA Details
Gene Symbol: Wbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053766.1 | NP_446218.1 | 10 | 1153 | ||
XM_008770840.1 | XP_008769062.1 | 7 | 1669 | |||
XM_008770841.1 | XP_008769063.1 | 8 | 1172 | |||
GenBank mRNA | BC089052.1 | - | 10 | 1153 | 7307299 |
