siRNA Details
| Gene Symbol: Ccl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057203.1 | NP_476551.1 | 3 | 1044 | ||
| GenBank mRNA | AF163477.1 | - | 3 | 1000 | ||
| BC088218.1 | - | 3 | 1044 | 7384271 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl22| Gene Symbol: Ccl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057203.1 | NP_476551.1 | 3 | 1044 | ||
| GenBank mRNA | AF163477.1 | - | 3 | 1000 | ||
| BC088218.1 | - | 3 | 1044 | 7384271 | ||