siRNA Details
Gene Symbol: Ccl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057203.1 | NP_476551.1 | 3 | 1086 | ||
GenBank mRNA | AF163477.1 | - | 3 | 1042 | ||
BC088218.1 | - | 3 | 1086 | 7384271 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057203.1 | NP_476551.1 | 3 | 1086 | ||
GenBank mRNA | AF163477.1 | - | 3 | 1042 | ||
BC088218.1 | - | 3 | 1086 | 7384271 |