siRNA Details
| Gene Symbol: Nexn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139230.1 | NP_631976.1 | 9 | 1397 | ||
| NM_139231.1 | NP_631977.1 | 9 | 1247 | |||
| XM_006233479.2 | XP_006233541.1 | 11 | 1565 | |||
| XM_006233480.2 | XP_006233542.1 | 11 | 1814 | |||
| XM_006233483.2 | XP_006233545.1 | 10 | 1523 | |||
| XM_006233484.2 | XP_006233546.1 | 10 | 1372 | |||
| XM_006233485.2 | XP_006233547.1 | 9 | 1330 | |||
| XM_008761548.1 | XP_008759770.1 | 10 | 1514 | |||
| GenBank mRNA | AF056034.1 | - | 9 | 1397 | ||
| AF056035.1 | - | 9 | 1247 | |||
| EU723599.1 | - | 4 | 784 | |||

Nexn
Genomic Map