siRNA Details
| Gene Symbol: ARID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152641.2 | NP_689854.2 | 15 | 2621 | ||
| XM_006719272.3 | XP_006719335.1 | 15 | 2775 | |||
| XM_011538025.1 | XP_011536327.1 | 9 | 1728 | |||
| GenBank mRNA | AL832200.1 | - | 11 | 2174 | ||
| AY727870.2 | - | 15 | 2621 | |||
| BX537879.1 | - | 1 | 249 | |||
| BX648152.1 | - | 9 | 1912 | |||
| BX649133.1 | - | 7 | 1467 | |||
| CR749833.1 | - | 14 | 2540 | |||
| DQ096628.1 | - | 15 | 2621 | |||

ARID2
Genomic Map