siRNA Details
| Gene Symbol: St8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013666.2 | NP_038694.2 | 8 | 1643 | ||
| NM_153124.2 | NP_694764.2 | 7 | 1535 | |||
| GenBank mRNA | AK013542.1 | - | 7 | 1532 | ||
| AK078670.1 | - | 7 | 1319 | |||
| AK162549.1 | - | 7 | 1315 | |||
| BC034855.1 | - | 7 | 1529 | 5146703 | ||
| X98014.1 | - | 8 | 1505 | |||

St8sia5
Genomic Map