siRNA Details
| Gene Symbol: Arhgef15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177566.3 | NP_808234.2 | 3 | 1043 | ||
| GenBank mRNA | BC089365.1 | - | 3 | 1043 | 6844583 | |
| BC116343.1 | - | 3 | 1019 | 40098168 | ||
| BC116344.1 | - | 3 | 1019 | 40098169 | ||

Arhgef15
Genomic Map