siRNA Details
| Gene Symbol: Thnsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001297.2 | NP_001001297.1 | 3 | 849 | ||
| NM_177588.2 | NP_808256.1 | 3 | 2421 | |||
| GenBank mRNA | AK029518.1 | - | 3 | 2422 | ||
| AK033465.1 | - | 3 | 818 | |||
| AK083895.1 | - | 3 | 2421 | |||
| AK087148.1 | - | 3 | 932 | |||
| BC027743.1 | - | 1 | 946 | |||
| BC090667.1 | - | 1 | 2130 | 30686917 | ||
| BC117854.2 | - | 2 | 2205 | 40094242 | ||

Thnsl1
Genomic Map