siRNA Details
| Gene Symbol: Olr1404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000787.1 | NP_001000787.1 | 1 | 636 | ||
| GenBank mRNA | X89699.1 | - | 1 | 262 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1404| Gene Symbol: Olr1404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000787.1 | NP_001000787.1 | 1 | 636 | ||
| GenBank mRNA | X89699.1 | - | 1 | 262 | ||