siRNA Details
| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001182.3 | NP_001001182.2 | 8 | 1345 | ||
| GenBank mRNA | AK029231.1 | - | 2 | 393 | ||
| AK030867.1 | - | 7 | 1261 | |||
| AK134250.1 | - | 9 | 1422 | |||
| AK147737.1 | - | 9 | 1537 | |||
| AK173180.1 | - | 9 | 1443 | |||
| BC027336.1 | - | 5 | 970 | |||
| BC150814.1 | - | 9 | 1327 | 9087725 | ||

Baz2b
Genomic Map