siRNA Details
| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001182.3 | NP_001001182.2 | 9 | 1623 | ||
| GenBank mRNA | AK029231.1 | - | 3 | 671 | ||
| AK030867.1 | - | 8 | 1539 | |||
| AK134250.1 | - | 10 | 1700 | |||
| AK147737.1 | - | 10 | 1815 | |||
| AK173180.1 | - | 10 | 1721 | |||
| BC027336.1 | - | 6 | 1248 | |||
| BC150814.1 | - | 10 | 1605 | 9087725 | ||

Baz2b
Genomic Map