siRNA Details
| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001182.3 | NP_001001182.2 | 36 | 7440 | ||
| GenBank mRNA | BC042646.1 | - | 3 | 1293 | ||
| BC053917.1 | - | 10 | 3096 | |||
| BC150814.1 | - | 38 | 7344 | 9087725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baz2b| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001182.3 | NP_001001182.2 | 36 | 7440 | ||
| GenBank mRNA | BC042646.1 | - | 3 | 1293 | ||
| BC053917.1 | - | 10 | 3096 | |||
| BC150814.1 | - | 38 | 7344 | 9087725 | ||