siRNA Details
| Gene Symbol: CLN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017882.2 | NP_060352.1 | 7 | 2048 | ||
| GenBank mRNA | AK000568.1 | - | 7 | 1989 | ||
| AK027604.1 | - | 7 | 2012 | |||
| AK074753.1 | - | 4 | 1538 | |||
| BC010849.1 | - | 7 | 1958 | 3878776 | ||
| BC013130.1 | - | 7 | 1895 | 4517748 | ||

CLN6
Genomic Map