siRNA Details
| Gene Symbol: Slpil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008872.1 | NP_001008872.1 | 2, 3 | 252 | ||
| XM_008762532.1 | XP_008760754.1 | 2, 3 | 252 | |||
| Gene Symbol: LOC103694893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773874.1 | XP_008772096.1 | 2, 3 | 252 | ||

Slpil2
Genomic Map