siRNA Details
| Gene Symbol: WDR13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000570.1 | - | 761 | |||
| AK225542.1 | - | 761 | ||||
| BC002507.2 | - | 1177 | ||||
| BX649101.1 | - | 1060 | ||||
| DQ907252.1 | - | 1807 | ||||
| DQ907255.1 | - | 2065 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR13| Gene Symbol: WDR13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000570.1 | - | 761 | |||
| AK225542.1 | - | 761 | ||||
| BC002507.2 | - | 1177 | ||||
| BX649101.1 | - | 1060 | ||||
| DQ907252.1 | - | 1807 | ||||
| DQ907255.1 | - | 2065 | ||||