siRNA Details
Gene Symbol: Trim40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009175.1 | NP_001009175.1 | 1, 2 | 346 | ||
XM_008772696.1 | XP_008770918.1 | 1, 2 | 346 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009175.1 | NP_001009175.1 | 1, 2 | 346 | ||
XM_008772696.1 | XP_008770918.1 | 1, 2 | 346 |