siRNA Details
| Gene Symbol: Vom1r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009517.1 | NP_001009517.1 | 1 | 60 | ||
| GenBank mRNA | AY510342.1 | - | 1 | 60 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r3| Gene Symbol: Vom1r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009517.1 | NP_001009517.1 | 1 | 60 | ||
| GenBank mRNA | AY510342.1 | - | 1 | 60 | ||