siRNA Details
| Gene Symbol: Clp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009599.1 | NP_001009599.1 | 2 | 262 | ||
| XM_006234461.2 | XP_006234523.1 | 2 | 797 | |||
| XM_006234462.1 | XP_006234524.1 | 2 | 345 | |||
| GenBank mRNA | BC087130.1 | - | 2 | 262 | 7112583 | |
| FQ231160.1 | - | 2 | 289 | |||

Clp1
Genomic Map