siRNA Details
| Gene Symbol: Psph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009679.1 | NP_001009679.1 | 4 | 543 | ||
| GenBank mRNA | BC088310.1 | - | 4 | 543 | 7324292 | |
| FQ213675.1 | - | 4 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psph| Gene Symbol: Psph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009679.1 | NP_001009679.1 | 4 | 543 | ||
| GenBank mRNA | BC088310.1 | - | 4 | 543 | 7324292 | |
| FQ213675.1 | - | 4 | 534 | |||