siRNA Details
| Gene Symbol: Hsd17b13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009684.1 | NP_001009684.1 | 3 | 491 | ||
| GenBank mRNA | BC088191.1 | - | 3 | 491 | 7372031 | |
| FQ218851.1 | - | 1 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd17b13| Gene Symbol: Hsd17b13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009684.1 | NP_001009684.1 | 3 | 491 | ||
| GenBank mRNA | BC088191.1 | - | 3 | 491 | 7372031 | |
| FQ218851.1 | - | 1 | 86 | |||