siRNA Details
| Gene Symbol: Dcaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009686.2 | NP_001009686.1 | 6 | 765 | ||
| XM_006251971.2 | XP_006252033.1 | 7 | 1100 | |||
| GenBank mRNA | BC081957.1 | - | 7 | 910 | ||
| BC087110.1 | - | 5 | 546 | 7193336 | ||
| BC091364.1 | - | 6 | 765 | 7319156 | ||
| FQ222851.1 | - | 7 | 1066 | |||

Dcaf11
Genomic Map