siRNA Details
| Gene Symbol: Rom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009690.1 | NP_001009690.1 | 3 | 1502 | ||
| GenBank mRNA | AY212508.1 | - | 3 | 1301 | ||
| BC087605.1 | - | 3 | 1502 | 7312188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rom1| Gene Symbol: Rom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009690.1 | NP_001009690.1 | 3 | 1502 | ||
| GenBank mRNA | AY212508.1 | - | 3 | 1301 | ||
| BC087605.1 | - | 3 | 1502 | 7312188 | ||