siRNA Details
| Gene Symbol: Dclk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009691.3 | NP_001009691.3 | 3 | 1424 | ||
| NM_001195832.1 | NP_001182761.1 | 3 | 1424 | |||
| XM_006232679.2 | XP_006232741.1 | 3 | 1594 | |||
| XM_006232680.2 | XP_006232742.1 | 3 | 1594 | |||
| XM_006232681.2 | XP_006232743.1 | 3 | 1593 | |||
| XM_006232682.2 | XP_006232744.1 | 3 | 1593 | |||
| XM_006232684.2 | XP_006232746.1 | 3 | 1594 | |||
| XM_006232685.2 | XP_006232747.1 | 3 | 1592 | |||
| XM_008761176.1 | XP_008759398.1 | 3 | 1595 | |||
| XM_008761177.1 | XP_008759399.1 | 3 | 489 | |||
| XM_008761178.1 | XP_008759400.1 | 3 | 1592 | |||
| GenBank mRNA | AY673997.1 | - | 3 | 816 | ||
| AY673998.1 | - | 3 | 896 | |||
| AY673999.1 | - | 1 | 12 | |||
| AY674000.1 | - | 3 | 816 | |||
| FM030581.1 | - | 2 | 160 | |||

Dclk2
Genomic Map