siRNA Details
Gene Symbol: Dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009696.1 | NP_001009696.1 | 5 | 616 | ||
GenBank mRNA | BC087627.1 | - | 5 | 616 | 7318159 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009696.1 | NP_001009696.1 | 5 | 616 | ||
GenBank mRNA | BC087627.1 | - | 5 | 616 | 7318159 |