siRNA Details
| Gene Symbol: Mtfmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009697.1 | NP_001009697.1 | 9 | 1099 | ||
| GenBank mRNA | BC088470.1 | - | 9 | 1099 | 7128152 | |
| FQ226710.1 | - | 5 | 426 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtfmt| Gene Symbol: Mtfmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009697.1 | NP_001009697.1 | 9 | 1099 | ||
| GenBank mRNA | BC088470.1 | - | 9 | 1099 | 7128152 | |
| FQ226710.1 | - | 5 | 426 | |||