siRNA Details
| Gene Symbol: Lysmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009698.1 | NP_001009698.1 | 3 | 937 | ||
| GenBank mRNA | BC088262.1 | - | 3 | 937 | 7376577 | |
| FQ225025.1 | - | 2 | 655 | |||
| FQ225894.1 | - | 2 | 655 | |||
| FQ227395.1 | - | 2 | 655 | |||
| FQ234752.1 | - | 2 | 657 | |||

Lysmd3
Genomic Map