siRNA Details
| Gene Symbol: Imp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009700.1 | NP_001009700.1 | 9 | 1251 | ||
| GenBank mRNA | BC087066.1 | - | 9 | 1251 | 7132412 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Imp4| Gene Symbol: Imp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009700.1 | NP_001009700.1 | 9 | 1251 | ||
| GenBank mRNA | BC087066.1 | - | 9 | 1251 | 7132412 | |