siRNA Details
Gene Symbol: Itfg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009701.1 | NP_001009701.1 | 486 | |||
GenBank mRNA | BC088404.1 | - | 486 | 7108395 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Itfg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009701.1 | NP_001009701.1 | 486 | |||
GenBank mRNA | BC088404.1 | - | 486 | 7108395 |