siRNA Details
| Gene Symbol: Rrp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009702.1 | NP_001009702.1 | 2 | 332 | ||
| GenBank mRNA | BC087649.1 | - | 2 | 332 | 7310706 | |
| FQ215627.1 | - | 2 | 331 | |||
| FQ219085.1 | - | 2 | 353 | |||
| FQ225526.1 | - | 2 | 356 | |||

Rrp15
Genomic Map