siRNA Details
| Gene Symbol: Pip5k1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009967.2 | NP_001009967.2 | 6 | 552 | ||
| NM_001033970.1 | NP_001029142.1 | 6 | 552 | |||
| XM_006240935.2 | XP_006240997.1 | 6 | 595 | |||
| XM_008765101.1 | XP_008763323.1 | 6 | 595 | |||
| XM_008765102.1 | XP_008763324.1 | 6 | 595 | |||
| XM_008765103.1 | XP_008763325.1 | 6 | 595 | |||
| GenBank mRNA | AY850259.1 | - | 6 | 491 | ||
| BC101909.1 | - | 6 | 524 | 7933923 | ||
| CB557931.1 | - | 6 | 563 | |||
| CK366332.1 | - | 6 | 541 | |||

Pip5k1c
Genomic Map