siRNA Details
| Gene Symbol: Pea15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY487415.1 | - | 87 | |||
| BC132661.1 | - | 184 | 40130938 | |||
| BC132663.1 | - | 184 | 40130940 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_001010832.1 | - | 87 | Chr. - - [-] on Build | ||

Pea15b
Genomic Map