siRNA Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010833.2 | NP_001010833.2 | 3 | 474 | ||
| GenBank mRNA | AK033009.1 | - | 3 | 311 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mdc1| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010833.2 | NP_001010833.2 | 3 | 474 | ||
| GenBank mRNA | AK033009.1 | - | 3 | 311 | ||