siRNA Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010833.2 | NP_001010833.2 | 15, 16 | 5152 | ||
| GenBank mRNA | AK129074.1 | - | 9, 10 | 2888 | ||
| BC085140.1 | - | 10, 11 | 3021 | |||
| BC094363.1 | - | 11, 12 | 3855 | |||

Mdc1
Genomic Map