siRNA Details
| Gene Symbol: Slc10a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010834.2 | NP_001010834.1 | 1 | 1283 | ||
| GenBank mRNA | AK143900.1 | - | 1 | 1284 | ||
| AY825925.1 | - | 1 | 1225 | |||
| BC141377.1 | - | 1 | 1260 | 9055990 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc10a5| Gene Symbol: Slc10a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010834.2 | NP_001010834.1 | 1 | 1283 | ||
| GenBank mRNA | AK143900.1 | - | 1 | 1284 | ||
| AY825925.1 | - | 1 | 1225 | |||
| BC141377.1 | - | 1 | 1260 | 9055990 | ||