siRNA Details
| Gene Symbol: CLVS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010852.3 | NP_001010852.2 | 3 | 1864 | ||
| XM_017010266.1 | XP_016865755.1 | 3 | 3566 | |||
| GenBank mRNA | AK095527.1 | - | 3 | 1868 | ||
| AK295158.1 | - | 3 | 326 | |||
| BC101376.1 | - | 3 | 1398 | 40025886 | ||
| BC101377.2 | - | 3 | 1400 | 40025889 | ||

CLVS2
Genomic Map