siRNA Details
| Gene Symbol: TTC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017904.3 | NP_060374.2 | 6 | 1561 | ||
| GenBank mRNA | AK000626.1 | - | 6 | 1491 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC22| Gene Symbol: TTC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017904.3 | NP_060374.2 | 6 | 1561 | ||
| GenBank mRNA | AK000626.1 | - | 6 | 1491 | ||