siRNA Details
| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027139.1 | 4, 5 | 869 | |||
| GenBank mRNA | AK123122.1 | - | 5, 6 | 1063 | ||
| BC018073.1 | - | 4, 5 | 813 | |||
| BC032490.1 | - | 4, 5 | 823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPIN3| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027139.1 | 4, 5 | 869 | |||
| GenBank mRNA | AK123122.1 | - | 5, 6 | 1063 | ||
| BC018073.1 | - | 4, 5 | 813 | |||
| BC032490.1 | - | 4, 5 | 823 | |||