siRNA Details
Gene Symbol: TMEM201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010866.3 | NP_001010866.1 | 6 | 3409 | ||
GenBank mRNA | BC121187.1 | - | 6 | 3364 | 3870153 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010866.3 | NP_001010866.1 | 6 | 3409 | ||
GenBank mRNA | BC121187.1 | - | 6 | 3364 | 3870153 |