siRNA Details
| Gene Symbol: SLC10A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010893.2 | NP_001010893.1 | 1 | 63 | ||
| GenBank mRNA | AK095808.1 | - | 1 | 1268 | ||
| AY825924.1 | - | 1 | 63 | |||
| BC136625.1 | - | 1 | 143 | 9020615 | ||
| BC136631.1 | - | 1 | 143 | 9020621 | ||

SLC10A5
Genomic Map