siRNA Details
| Gene Symbol: Slc25a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010958.1 | NP_001010958.1 | 4 | 1179 | ||
| XM_006240541.2 | XP_006240603.1 | 4 | 1340 | |||
| GenBank mRNA | BC089065.1 | - | 4 | 1179 | 7319298 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a29| Gene Symbol: Slc25a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010958.1 | NP_001010958.1 | 4 | 1179 | ||
| XM_006240541.2 | XP_006240603.1 | 4 | 1340 | |||
| GenBank mRNA | BC089065.1 | - | 4 | 1179 | 7319298 | |