siRNA Details
| Gene Symbol: Olfr332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011770.2 | NP_001011770.2 | 2 | 1037 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr332| Gene Symbol: Olfr332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011770.2 | NP_001011770.2 | 2 | 1037 | ||